Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol, 99.78%, MP Biomedicals™

Catalog No. ICN19015001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19015001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19015001 Supplier MP Biomedicals, Inc Supplier No. 0219015001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 315-30-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00599413
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
PubChem CID 2094
ChEBI CHEBI:40279
IUPAC Name 1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2NNC=C12

Specifications

Color White
Assay Percent Range 0.9978
Quantity 1 g
Formula Weight 136.11
Physical Form Powder
Chemical Name or Material Allopurinol
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.