Learn More
Selleck Chemical LLC TCS7010 (Aurora A Inhibitor I) S1451-5mg
SDP
Supplier:  Selleck Chemical LLC S14515MG
Encompass_Preferred
TCS7010 (Aurora A Inhibitor I) is a novel potent and selective inhibitor of Aurora A with IC50 of 3 4 nM in a cell-free assay It is 1000-fold more selective for Aurora A than Aurora B Aurora A Inhibitor I (TC-S 7010) triggers apoptosis through the ROS-mediated UPR signaling pathway
Catalog No. 50-310-8686
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.