Learn More
Selleck Chemical LLC Oxybutynin S1754-1g
SDP
Supplier:  Selleck Chemical LLC S17541G
Encompass_Preferred
Oxybutynin is a competitive antagonist of the M1 M2 and M3 subtypes of the muscarinic acetylcholine receptor used to relieve urinary and bladder difficulties
Catalog No. 50-311-0127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.