Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600407 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-407 500 g Amber Glass Bottle
18-600-404 5 g Amber Glass Bottle
18-600-405 25 g Amber Glass Bottle
18-600-408 100 g Amber Glass Bottle
18-600-409 1 kg Amber Glass Bottle
18-600-406 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-407 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1577500GMBL
Only null left
Add to Cart
Add to Cart
Request a Quote

A1577, 315-30-0, C5H4N4O

Spectrum™ Chemical Allopurinol, USP is a xanthine oxidase inhibitor that is primarily used to treat Hyperuricemia. It is on the World Health Organization′s List of Essential Medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 315-30-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00599413
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
IUPAC Name 1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2NNC=C12

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.