Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1
Catalog No. 02215115 Shop All Troemner Products
Change view
Click to view available options
Quantity:
12/Set
17/Set
21/Set
22/Set
23/Set
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-215-115 22/Set
02-216-016 12/Set
02-319B 17/Set
02-215-120 23/Set
02-319D 21/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-215-115 Supplier Troemner™ Supplier No. 30391341
Add to Cart
Add to Cart
Includes
Forceps
Excellent for high-precision calibrations and reference standards
Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2 (2mg, 20mg, etc.) and one weight for all other inclusive values
  • Certificate of calibration is available; contact your Fisher Customer Service Representative
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medium Metric Analytical Weight Set
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 1 mg to 200 g
Mass 200 g, 100 g, 50 g, (2)20 g, 10 g, 5 g, (2)2 g, 1 g, 500 mg, (2)200 mg, 100 mg, 50 mg, (2)20 mg, 10 mg, 5 mg, (2)2 mg, 1 mg
Includes Stainless Steel cylindrical weight: 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminum sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Quantity 22/Set
Packaging Quantity Multiple