Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1 with Traceable Certificate
Catalog No. 02216017 Shop All Troemner Products
Change view
Click to view available options
Quantity:
12/Set
14/Set
16/Set
17/Set
21/Set
22/Set
23/Set
24/Set
8/Set
9/Set
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-216-017 12/Set
02-216-004 24/Set
02-319DT 21/Set
02-319HT 16/Set
02-215-115T 22/Set
02-319AT 9/Set
02-319-GT 8/Set
02-319BT 17/Set
02-216-013 14/Set
02-100-179 23/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-216-017 Supplier Troemner™ Supplier No. 30391538
Add to Cart
Add to Cart
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weights sets 1mg-500g are provided in a high quality polycarbonate set case
  • Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
  • Each polycarbonate case is placed in its own individual compartment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Small Metric Analytical Weight Set
Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 1 to 500 mg
Mass 500 mg, (2)200 mg, 100 mg, 50 mg, (2)20 mg, 10 mg, 5 mg, (2)2 mg, 1 mg
Includes Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminum sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Quantity 12/Set
Packaging Quantity Multiple