Learn More
Grace™ Alltech™ Heliflex™ AT™-1701 Capillary Columns
Catalog No. AT13417
Change view
Click to view available options
Length (Metric):
30 m
Catalog No. Length (Metric)
AT13417 30 m
1 options
Catalog No. AT13417 Supplier Grace™ Supplier No. 13417
Similar to DB-1701, HP-1701, OV-1701
  • (14% cyanopropylphenyl)-86% methylpolysiloxan
  • Intermediate polarity columns are manufactured from (14% cyanopropylphenyl)-86% methylpolysiloxane (polyamide-coated synthetic fused silica)
  • Mounted on a rugged wear-resistant cage
  • Individually tested to guarantee performance
  • Include a test chromatogram, instruction manual, capillary end caps, and ceramic column cutter
Specifications
Length (Metric) 30 m