Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl alcohol, 98+%

Catalog No. AAA15026AP
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

Catalog No. AAA15026AP

Supplier: Thermo Scientific Chemicals A15026AP

Only null left
Add to Cart

CAS: 107-18-6 | C3H6O | 58.08 g/mol

Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, oleum and alkali metals. Polymerizes during prolonged storage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-18-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00002920
InChI Key XXROGKLTLUQVRX-UHFFFAOYSA-N
Synonym allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench
PubChem CID 7858
ChEBI CHEBI:16605
IUPAC Name prop-2-en-1-ol
SMILES OCC=C

Specifications

Melting Point −129°C
Density 0.852
Boiling Point 95°C to 97°C
Flash Point 22°C (71°F)
Assay Percent Range ≥98%
Refractive Index 1.413
Linear Formula CH2=CHCH2OH
Quantity 500 mL
UN Number UN1098
Beilstein 605307
Merck Index 14,286
Solubility Information Miscible with water.
Formula Weight 58.08
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Allyl Alcohol
Show More Show Less
Hazard Category H225-H301-H310+H330-H315-H319-H335
Hazard Statement GHS H Statement
H225-H301-H311-H331-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYL ALCOHOL
EINECSNumber 203-470-7
RTECSNumber BA5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.