Learn More
Allyl alcohol, 98+%
Description
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, acids, oleum and alkali metals. Polymerizes during prolonged storage.
Specifications
Specifications
| Melting Point | -129°C |
| Density | 0.852 |
| Boiling Point | 95°C to 97°C |
| Flash Point | 22°C (71°F) |
| Odor | Pungent |
| Assay Percent Range | ≥98% |
| Refractive Index | 1.413 |
| Linear Formula | CH2=CHCH2OH |
| Quantity | 2500 mL |
| UN Number | UN1098 |
| Show More |
Safety and Handling
| Hazard Category | H225-H301-H310+H330-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H301-H311-H331-H315-H319-H335 Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYL ALCOHOL |
| EINECSNumber | 203-470-7 |
| RTECSNumber | BA5075000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.