Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Benzyl Ether 99.0+%, TCI America™
SDP

Catalog No. A1427100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1427100ML 100 mL
A142725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1427100ML Supplier TCI America Supplier No. A1427100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14593-43-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00078288
InChI Key HUGHWHMUUQNACD-UHFFFAOYSA-N
PubChem CID 84542
IUPAC Name prop-2-enoxymethylbenzene
SMILES C=CCOCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 205°C
Quantity 100 mL
Formula Weight 148.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Benzyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.