Learn More
Sigma Organic Chemistry Allyl bromide | 1KG | 106-95-6 | MFCD00000244
Supplier: Sigma Organic Chemistry A295851KG

Allyl bromide, 1KG
About This Item:
Synonym(s): 3-Bromo-1-propene
Linear Formula: CH2=CHCH2Br
Molecular Weight: 120.98
CAS Number: 106-95-6
MDL Number: MFCD00000244
UNSPSC Code: 12352101
Purity: 0.99
Boiling Point: 70-71 C (lit.)
Melting Point: -119 C (lit.)
Density: 1.398 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.469 (lit.)
Storage: 2-8C
EINECS Number: 203-446-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.