Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Cyclohexanepropionate, Spectrum™ Chemical
SDP

Catalog No. 18620107 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-107 25 mL
18-620-108 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-107 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A231525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2705-87-5
Molecular Weight (g/mol) 196.29

Specifications

Quantity 25 mL
Packaging Amber Glass
Boiling Point 91°C
Percent Purity 98%
Grade Ungraded
Chemical Name or Material Allyl Cyclohexanepropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.