Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl ethyl ether, 98%, Thermo Scientific™

Catalog No. AAB2227618
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2227618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2227618 Supplier Thermo Scientific Chemicals Supplier No. B2227618

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Allyl ethyl ether is used as an intermediate to manufacture other chemicals.

Solubility
Soluble in organic solvents, miscible with ethanol, diethyl ether. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-31-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009255
InChI Key OJPSFJLSZZTSDF-UHFFFAOYSA-N
Synonym allyl ethyl ether, 1-propene, 3-ethoxy, allylethylether, ethyl allyl ether, unii-sm8t658re3, ether, allyl ethyl, 3-ethoxy-1-propene, ethyl 2-propenyl ether, 3-ethoxypropene, acmc-1atoi
PubChem CID 11191
IUPAC Name 3-ethoxyprop-1-ene
SMILES CCOCC=C

Specifications

Density 0.76
Boiling Point 64°C to 65°C
Flash Point −20°C (−4°F)
Refractive Index 1.388
Linear Formula CH2=CHCH2OCH2CH3
Quantity 50 g
UN Number UN2335
Beilstein 1361480
Sensitivity Air sensitive
Merck Index 14,293
Solubility Information Soluble in organic solvents,miscible with ethanol,diethyl ether. Insoluble in water.
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material Allyl ethyl ether
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H302-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL ETHYL ETHER
EINECSNumber 209-169-7
RTECSNumber KM9120000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.