Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol

Catalog No. ac102950050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102950050 5 g Glass bottle
AC102951000 100 g Glass bottle
AC102955000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102950050 Supplier Thermo Scientific Chemicals Supplier No. 102950050
Only null left
Add to Cart

CAS: 57-06-7 | C4H5NS | 99.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-06-7
MDL Number MFCD00004822
InChI Key ZOJBYZNEUISWFT-UHFFFAOYSA-N
Synonym allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
PubChem CID 5971
ChEBI CHEBI:73224
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point -80.0°C
Color White
Density 1.0175g/mL
Boiling Point 151.0°C to 153.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5NS
Linear Formula H2C=CHCH2NCS
Refractive Index 1.5290 to 1.5310
Quantity 5 g
Beilstein 04, 214
Specific Gravity 1.0175
Merck Index 15, 286
Solubility Information Solubility in water: 2g/L water (20°C). Other solubilities: ethanol of 65 vol%- 1:20 v/v,diethylphthalate - in all proportions,paraffin oil 65 cp- in all proportions,1,2-propanediol - 1:5 v/v,soluble in ether,acetone,benzene,cs2
Molecular Weight (g/mol) 99.15
Stabilizer 0.01% alpha-tocopherol
Formula Weight 99.15
Percent Purity 94%
Physical Form Fine Crystalline Powder
Chemical Name or Material Allyl isothiocyanate, 94%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very t
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov
EINECSNumber 200-309-2
RTECSNumber NX8225000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.