Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl methyl carbonate, 98%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
35466-83-2
C5H8O3
116.12
MFCD00134545
YHLVIDQQTOMBGN-UHFFFAOYSA-N
allyl methyl carbonate,methyl prop-2-en-1-yl carbonate,carbonic acid allyl methyl ester,isobutylene carbonate,methyl allyl carbonate,acmc-1agon,allyl methyl carbonate #,prop-2-enyl methoxyformate,yhlvidqqtombgn-uhfffaoysa
534788
methyl prop-2-enyl carbonate
COC(=O)OCC=C
35466-83-2
C5H8O3
116.12
MFCD00134545
YHLVIDQQTOMBGN-UHFFFAOYSA-N
allyl methyl carbonate,methyl prop-2-en-1-yl carbonate,carbonic acid allyl methyl ester,isobutylene carbonate,methyl allyl carbonate,acmc-1agon,allyl methyl carbonate #,prop-2-enyl methoxyformate,yhlvidqqtombgn-uhfffaoysa
534788
methyl prop-2-enyl carbonate
COC(=O)OCC=C
Specific Gravity 1.022
Flash Point 33°C
Boiling Point 131.0°C
Quantity 25mL
Color Colorless
Density 1.0220g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH2=CHCH2OCO2CH3
Formula Weight 116.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Allyl methyl carbonate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning