Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Methyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. A111725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A111725ML 25 mL
A11175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A111725ML Supplier TCI America Supplier No. A111725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10152-76-8
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00008657
InChI Key NVLPQIPTCCLBEU-UHFFFAOYSA-N
Synonym allyl methyl sulfide, 1-propene, 3-methylthio, allyl methyl sulphide, 3-methylthio-1-propene, sulfide, allyl methyl, methyl propenyl sulfide, unii-v7qi1r316c, methyl allyl sulfide, ccris 7066
PubChem CID 66282
IUPAC Name 3-(methylsulfanyl)prop-1-ene
SMILES CSCC=C

Specifications

Color Colorless
Boiling Point 92°C
Quantity 25 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Sulfide
EINECSNumber (2)-0468
RTECSNumber UD1015000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.