Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Phenyl Sulfone 95.0+%, TCI America™
SDP

Catalog No. A14445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14445G Supplier TCI America Supplier No. A14445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 16212-05-8
Color Yellow
Boiling Point 128°C
Molecular Formula C9H10O2S
Quantity 5 g
MDL Number MFCD00014739
Synonym allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
InChI Key KYPIULIVYSQNNT-UHFFFAOYSA-N
SMILES C=CCS(=O)(=O)C1=CC=CC=C1
IUPAC Name prop-2-enylsulfonylbenzene
Molecular Weight (g/mol) 182.237
PubChem CID 85330
Formula Weight 182.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenyl Sulfone
Show More Show Less
RTECSNumber WR2400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.