Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylpentafluorobenzene 98.0+%, TCI America™
SDP

Catalog No. A161125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A161125G 25 g
A16115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A161125G Supplier TCI America Supplier No. A161125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1736-60-3
Molecular Formula C9H5F5
Molecular Weight (g/mol) 208.13
MDL Number MFCD00000301
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
PubChem CID 74435
IUPAC Name 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F

Specifications

Color Yellow
Boiling Point 149°C
Quantity 25 g
UN Number 1993
Formula Weight 208.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylpentafluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.