Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrimethoxysilane 97.0+%, TCI America™
SDP

Catalog No. A150425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A150425ML 25 mL
A15045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A150425ML Supplier TCI America Supplier No. A150425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2551-83-9
Molecular Formula C6H14O3Si
Molecular Weight (g/mol) 162.26
MDL Number MFCD00053865
InChI Key LFRDHGNFBLIJIY-UHFFFAOYSA-N
Synonym allyltrimethoxysilane, silane, trimethoxy-2-propenyl, silane, trimethoxy-2-propen-1-yl, allyl trimethoxy silane, silane, allyltrimethoxy, allytrimethoxysilane, acmc-209gkn, trimethoxy prop-2-enyl silane, silane,trimethoxy-2-propen-1-yl
PubChem CID 75698
IUPAC Name trimethoxy(prop-2-enyl)silane
SMILES CO[Si](CC=C)(OC)OC

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 mL
UN Number 1993
Formula Weight 162.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltrimethoxysilane
EINECSNumber (2)-2066
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.