Learn More
Medchemexpress LLC Almitrine mesylate | 29608-49-9 | MFCD01679060 | 99.8% | 669.76 g/mol | C28H37F2N7O6S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10731950MG
Encompass_Preferred
Almitrine mesylate is the methanesulfonate salt of almitrine, a peripheral chemoreceptor agonist that selectively inhibits Ca2+-dependent K+ channels. Supplied as a high-purity solid for research use, it is used in pharmacology studies investigating respiratory stimulation and potassium-channel modulation. Solubility and storage guidance facilitate formulation for in vitro and in vivo experiments.
  • Peripheral chemoreceptor agonist that modulates respiratory drive.
  • Selectively inhibits Ca2+-dependent K+ channels.
  • High purity suitable for research applications.
  • Soluble in DMSO; in vivo formulations reported for dosing.
  • Stable when stored sealed at 4°C; solution storage guidance available.
Catalog No. 50-002-40599