Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Alnico Bar Magnets
SDPSCI_ED

Catalog No. S15704 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W x H):
1.5 x 0.5 x .25 in.
2 x 0.5 x .25 in.
3 x 0.5 x .25 in.
4 x 0.5 x .25 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15704 2 x 0.5 x .25 in.
S15705 1.5 x 0.5 x .25 in.
S15862 3 x 0.5 x .25 in.
S15773 4 x 0.5 x .25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S15704 Supplier United Scientific™ Supplier No. ABM020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Alnico Bar Magnets are powerful magnets for classroom experiments.

  • Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers.
  • Magnets are painted with North and South poles, clearly marked.
  • Magnets available in five different sizes.

Specifications

Material Alnico
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnet
Dimensions (L x W x H) 2 x 0.5 x .25 in.

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.