Learn More
Description
- Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers.
- Magnets are painted with North and South poles, clearly marked.
- Magnets available in five different sizes.
Specifications
Specifications
| Material | Brass Wire |
| For Use With (Application) | Magnetic Demonstrations |
| Grade | 5+ |
| Product Type | Magnet |
| Dimensions (L x W x H) | 4 x 0.5 x .25 in. |
Warning: Cancer and reproductive harm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

