Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aloe Vera, 200:1, Powder, Spectrum™ Chemical
SDP

Catalog No. 18600429 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-429 500 g Amber Glass Bottle
18-600-427 25 g Amber Glass Bottle
18-600-428 100 g Amber Glass Bottle
18-600-431 12 kg Poly Pail
18-600-430 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-429 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1861500GM
Add to Cart
Add to Cart

A1861, 85507-69-3

Spectrum™ Chemical Aloe Vera, 200:1, Powder - Spectrum’s freeze dried aloe vera powder is easy to use. Just add 199 parts distilled or deionized water to 1 part aloe vera powder to reconstitute the oil derived from the Aloe Vera plant. It is known for its moisturizing and healing qualities and can be used in a wide range of personal care products. It is also a humectant, hair conditioners, and to relieve dandruff and dry scalp. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 85507-69-3

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.