Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX12 Mouse anti-Human, Clone: OTI1C3, liquid, TrueMAB™
SDP

Catalog No. 501681717
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1717 Supplier OriGene Supplier No. TA807718
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oxygenase and 14,15-leukotriene A4 synthase activity.

Specifications

Antigen ALOX12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALOX12
Gene Accession No. P18054
Gene Alias 12-LOX, 12S-LOX, LOG12
Gene Symbols ALOX12
Host Species Mouse
Immunogen Full length human recombit protein of human ALOX12 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.