Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX15 Monoclonal Antibody (OTI3G8)
GREENER_CHOICE

Catalog No. PIMA525853 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525853 Supplier Invitrogen™ Supplier No. MA525853
  • $709.50
    $541.65 / Each of 1

    Save $167.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

ALOX15 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Alox15
Gene Accession No. P16050, Q02759
Gene Alias 12/15-lipoxygenase; 12/15-LO; 12/15-LOX; 12-LO; 12-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX-1; 15LOX-1; 15-LOX2; Alox12; Alox12l; Alox15; Arachidonate 12-lipoxygenase, leukocyte-type; arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; L-12LO; LOG15
Gene Symbols Alox15
Host Species Mouse
Immunogen Full length human recombinant protein of ALOX15 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246, 81639
Target Species Human, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.