Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ALOX15 Recombinant Protein

Catalog No. 89009872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-009-872 10 μg
89-009-873 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-009-872 Supplier Abnova™ Supplier No. H00000246P0110
Add to Cart
Add to Cart

  • Molecular weight: 98.56kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification:glutathione sepharose 4 fast flow
  • Storage buffer:50 mM Tris-HCI, 10 mM reduced glutathione
  • pH=8.0 in elution buffer

Sequence: MGLYRIRVSTGASLYAGSNNQVQLWLVGQHGEAALGKRLWPARGKETELKVEVPEYLGPLLFVKLRKRHLLKDDAWFCNWISVQGPGAGDEVRFPCYRWVEGNGVLSLPEGTGRTVGEDPQGLFQKHREEELEERRKLYRWGNWKDGLILNMAGAKLYDLPVDERFLEDKRVDFEVSLAKGLADL
AIKDSLNVLTCWKDLDDFNRIFWCGQSKLAERVRDSWKEDALFGYQFLNGANPVVLRRSAHLPARLVFPPGMEELQAQLEKELEGGTLFEADFSLLDGIKANVILCSQQHLAAPLVMLKLQPDGKLLPMVIQLQLPRTGSPPPPLFLPTDPPMAWLLAKCWVRSSDFQLHELQSHLLRGHLMAEVIVVATMRCLPSIHPIFKLIIPHLRYTLEINVRARTGLVSDMGIFDQIMSTGGGGHVQLLKQAGAFLTYSSFCPPDDLADRGLLGVKSSFYPQDALRLWEIIYRYVEGIVSLHYKTDVAVKDDPELQTWCREITEIGLQGAQDRGFPVSLQARDQVCHFVTMCIFTCTGQHASVHLGQLDWYSWVPNAPCTMRLPPPTTKDATLETVMATLPNFHQASLQMSITWQLGRRQPVMVAVGQHEEEYFSGPEPKAVLKKFREELAALDKEIEIRNAKLDMPYEYLRPSVVENSVAI

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH29032
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 246
Molecular Weight (g/mol) 98.56
Name ALOX15 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias 15-LOX-1
Common Name ALOX15
Gene Symbol ALOX15
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.