Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX15B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA564932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564932 Supplier Thermo Scientific Supplier No. PA564932
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRAGTWDKVSV SIVGTRGESP PLPLDNLGKE FTAGAEEDFQ VTLPEDVGRV LLLRVHKAPP VLPLLGPLAP DAWFCRWFQL TPPRGGHLLF PCYQWLEGAG TLVLQEGTAK VSWAD Highest antigen sequence identity to the following orthologs - mouse 73%, rat 73%.

ALOX15B Polyclonal antibody specifically detects ALOX15B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX15B
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALOX15B
Gene Accession No. O15296
Gene Alias 15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; 8S-LOX; ALOX15B; Alox8; arachidonate 15-lipoxygenase 2; arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate 15-lipoxygenase, type B; arachidonate 8-lipoxygenase; arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; linoleate 13-lipoxygenase 15-LOb
Gene Symbols ALOX15B
Host Species Rabbit
Immunogen Recombinant Human ALOX15B
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.