Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Alp Assay Buffer, 100 mL

Catalog No. 502582478
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2478 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2478 Supplier Abcam Supplier No. AB171729100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized buffer for alkaline phosphatase activity assays.

  • Designed for use in ALP assays
  • Cited in 1 publication

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Wash Solution
Format Bottle
Target Molecule PPB1
Quantity 100 mL
For Use With (Application) PPB1 measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.