Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Alp Enzyme, Quantitative Assay Kit, 1 Unit

Catalog No. 502707014
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7014 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7014 Supplier Abcam Supplier No. AB1346781UNIT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Quantitative enzyme assay kit cited in scientific literature.

  • Quantitative assay kit
  • Cited in 1 publication

Specifications

Kit Contents ALP Enzyme
Label or Dye Alkaline Phosphatase
Reactivity Alkaline Phosphatase
Target Molecule Alkaline Phosphatase
Quantity 1 Unit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.