Learn More
Selleck Chemical LLC Alpelisib (BYL719) S2814-1g
SDP
Supplier:  Selleck Chemical LLC S28141G
Encompass_Preferred
Alpelisib (BYL719) is a potent and selective PI3K inhibitor with IC50 of 5 nM in a cell-free assay and minimal effect on PI3K / / Phase 2
Catalog No. 50-310-8125
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.