Learn More
Selleck Chemical LLC Alpelisib (BYL719) 10mM/1mL 1217486-61-7
SDP
Supplier:  Selleck Chemical LLC S281410MM/1ML
Encompass_Preferred
Alpelisib (BYL719) is a potent and selective PI3K? inhibitor with IC50 of 5 nM in a cell-free assay, and minimal effect on PI3K?/?/?. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3003
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.