Learn More
Cellular Engineering Technologies Cryopreserved vial of induced pluripotent stem cells from a single female donor with Alpha 1 antitrypsin deficiency (500,000 cells)
Supplier:  Cellular Engineering Technologies CR1014500
Encompass_Preferred
Cell line was derived from a 57-year-old female donor with two copies of the SERPINA1 gene that produces an abnormal type of Alpha-1 protein. Donor cells were isolated and then programmed using our patented virus-free method using episomal DNA and our proprietary mix of vectors, excluding Myc and Lin28 transcription factors. Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753054