Learn More
Diagnostic Biosystems ALPHA-1-ANTITRYPSIN Alpha (α )-1-Antitrypsin
Supplier:  Diagnostic Biosystems RP04805
Encompass_Preferred
The antibody isspecific against human α-1-antitrypsin. This antibody stains macrophages in human tissues and does not stain any other cell type.
Catalog No. 50-238-5719
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.