Learn More
Biocare Medical, LLC Alpha-1-Fetoprotein (AFP) Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CP028A
Encompass_Preferred
This antibody reacts with human Alpha-1-Fetoprotein (AFP). Alpha-1-Fetoprotein antibody reacts with germ-cell tumors, gonadal tumors and liver carcinoma. Neoplasms commonly associated with Alpha-1-Fetoprotein (AFP) production are hepatocellular carcinomas and some germ cell tumors, typically yolk sac tumor. Rare tumors of visceral origin may also be associated with Alpha-1-Fetoprotein (AFP) production. Studies show that in hepatocellular carcinoma, Alpha-1-Fetoprotein (AFP) expression usually indicates malignancy in a hepatocellular nodule and hepatocytic histogenesis of a malignancy. Traces of contaminating antibodies have been removed by solid-phase absorption with human plasma proteins.
Catalog No. 50-827-49
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.