Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-1d Adrenoceptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575925 Supplier Invitrogen™ Supplier No. PA575925
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1d Adrenoceptor
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Adra1d
Gene Accession No. P25100
Gene Alias [a]1d; ADA1D; Adra1; Adra-1; Adra1a; Adra1d; ADRA1R; Adrd1; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; adrenoceptor alpha 1D; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; ALPHA1; alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; alpha1D-AR; Alpha1delta-AR; alpha-adrenergic receptor 1a; DAR; dJ779E11.2; Gpcr8; RA42; Spr8
Gene Symbols Adra1d
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids456-501 of Human alpha-1d Adrenoceptor.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.