Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-[2-(Methylamino)ethyl]benzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. M196025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M196025G 25 g
M19605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M196025G Supplier TCI America Supplier No. M196025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42142-52-9
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00674078
InChI Key XXSDCGNHLFVSET-UHFFFAOYSA-N
Synonym 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol
PubChem CID 2733989
IUPAC Name 3-(methylamino)-1-phenylpropan-1-ol
SMILES CNCCC(C1=CC=CC=C1)O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-[2-(Methylamino)ethyl]benzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.