Learn More
Invitrogen™ alpha-2a Adrenergic Receptor Polyclonal Antibody

Description
Positive test controls include: K-562, Mouse brain, Mouse Spinal cord. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: RRTRVPPSRR GPDAVAAPPG GTERRPNGLG PERSAGPGGA EAEPLPTQLN GAPGEPAPAG PRDTDALDLE ESSSSDHAER PPGPRRPERG PRGKGKARAS QVKPGDSLPR RGPGATGIGT PAAGPGEERV GAAKASRWRG R.
Specifications
Specifications
| Antigen | alpha-2a Adrenergic Receptor |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Adra2a |
| Gene Accession No. | P08913, P22909, Q01338 |
| Gene Alias | ADRA2; Adra-2; ADRA2A; Adra-2a; ADRA2R; ADRAR; adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; adrenoceptor alpha 2A; alpha(2A)AR; alpha-2 adrenergic receptor subtype C10; alpha2A; alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha2A-adrenergic receptor; ALPHA2AAR; alpha-2AAR; alpha2A-AR; alpha-2AAR subtype C10; alpha-2-adrenergic receptor, platelet type; alpha2-C10; alpha-2D adrenergic receptor; AW122659; CA2-47; OTTHUMP00000058849; RATRG20; RG20; subtype alpha2-C10; ZNF32 |
| Gene Symbols | Adra2a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.