Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-2a Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1048 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1048 Supplier Invitrogen™ Supplier No. PA1048
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-048 detects alpha-2A adrenergic receptor (A2AAR) from human, rat and mouse tissues. PA1-048 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~45 kDa protein representing A2AAR from mouse kidney membrane preparations. The PA1-048 immunogen is a synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR. This sequence is completely conserved between human, mouse, rat, and porcine A2AAR.

Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2a Adrenergic Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene Adra2a
Gene Accession No. P08913, P22909, Q01338
Gene Alias ADRA2; Adra-2; ADRA2A; Adra-2a; ADRA2R; ADRAR; adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; adrenoceptor alpha 2A; alpha(2A)AR; alpha-2 adrenergic receptor subtype C10; alpha2A; alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha2A-adrenergic receptor; ALPHA2AAR; alpha-2AAR; alpha2A-AR; alpha-2AAR subtype C10; alpha-2-adrenergic receptor, platelet type; alpha2-C10; alpha-2D adrenergic receptor; AW122659; CA2-47; OTTHUMP00000058849; RATRG20; RG20; subtype alpha2-C10; ZNF32
Gene Symbols Adra2a
Host Species Rabbit
Immunogen Synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11551, 150, 25083
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.