Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha-2a Adrenergic Receptor Recombinant Rabbit Monoclonal Antibody (3H29L10)

Catalog No. 703636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-363-6 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-363-6 Supplier Invitrogen™ Supplier No. 703636
  • $558.00
    $418.65 / Each of 1

    Save $139.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Cat, Pig, Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-2a Adrenergic Receptor
Applications Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3H29L10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Adra2a
Gene Accession No. P08913
Gene Alias ADRA2; Adra-2; ADRA2A; Adra-2a; ADRA2R; ADRAR; adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; adrenoceptor alpha 2A; alpha(2A)AR; alpha-2 adrenergic receptor subtype C10; alpha2A; alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha2A-adrenergic receptor; ALPHA2AAR; alpha-2AAR; alpha2A-AR; alpha-2AAR subtype C10; alpha-2-adrenergic receptor, platelet type; alpha2-C10; alpha-2D adrenergic receptor; AW122659; CA2-47; OTTHUMP00000058849; RATRG20; RG20; subtype alpha2-C10; ZNF32
Gene Symbols Adra2a
Host Species Rabbit
Immunogen Peptides corresponding to human Alpha-2a Adrenergic Receptor [1) aa225-aa238 2) aa180-aa194].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.