Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-(4-Piperidyl)benzhydrol 99.0+%, TCI America™
SDP

Catalog No. P136925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P136925G 25 g
P13695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P136925G Supplier TCI America Supplier No. P136925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115-46-8
Molecular Formula C18H21NO
Molecular Weight (g/mol) 267.372
MDL Number MFCD00066980
InChI Key ZMISODWVFHHWNR-UHFFFAOYSA-N
Synonym azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
PubChem CID 15723
IUPAC Name diphenyl(piperidin-4-yl)methanol
SMILES C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 267.37
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-(4-Piperidyl)benzhydrol
RTECSNumber TN0470000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.