Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7088471 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6813 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6813 Supplier Proteintech Group Inc Supplier No. 110321AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen alpha Actin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACTA1
Gene Accession No. P68032, P68033, P68035, P68133, P68134, P68136
Gene Alias ACTC, ACTC1, ACTC1 cardiac muscle 1, Actin, alpha cardiac muscle 1, actin, alpha, cardiac muscle 1, Alpha actin, Alpha cardiac actin, CMD1R, CMH11
Gene Symbols ACTA1, ACTC1
Host Species Rabbit
Immunogen alpha Actin Fusion Protein Ag1471
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11459, 11464, 29275, 29437, 58, 70
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.