Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Actinin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517308 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517308 Supplier Thermo Scientific Supplier No. PA517308
  • $722.00
    $547.65 / Each of 1

    Save $174.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

alpha Actinin 1 Polyclonal Antibody

alpha Actinin 1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene ACTN1
Gene Accession No. P12814, Q7TPR4, Q9Z1P2
Gene Alias 3110023F10Rik; actinin; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; Actn1; Actn1a; alpha actinin 1a; alpha-actinin; alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; fibroblast alpha actinin; FLJ40884; FLJ54432; hypothetical protein; hypothetical protein LOC524770; I79_004057; non-muscle alpha-actinin 1; non-muscle alpha-actinin-1
Gene Symbols ACTN1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to carboxy-terminal residues of human a-actinin-1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100755831, 109711, 81634, 87
Target Species Human, Mouse, Rat, Monkey, Hamster
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.