Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-m-xylene 96.0+%, TCI America™
SDP

Catalog No. T005110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005110G 10 g
T005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T005110G Supplier TCI America Supplier No. T005110G

Chemical Identifiers

CAS 36323-28-1
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00000132
InChI Key ZMCUKNMLHBAGMS-UHFFFAOYSA-N
Synonym 1,3-Bis(dibromomethyl)benzene, m-Xylylene Tetrabromide
PubChem CID 37418
IUPAC Name 1,3-bis(dibromomethyl)benzene
SMILES C1=CC(=CC(=C1)C(Br)Br)C(Br)Br

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 10 g
UN Number 1759
Formula Weight 421.75
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetrabromo-m-xylene
RTECSNumber ZE5150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.