Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-p-xylene 98.0+%, TCI America™
SDP

Catalog No. T005325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005325G 25 g
T00535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T005325G Supplier TCI America Supplier No. T005325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1592-31-0
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00017857
InChI Key VIQBABDKNOOCQD-UHFFFAOYSA-N
Synonym 1,4-Bis(dibromomethyl)benzene, p-Xylylene Tetrabromide
PubChem CID 74127
IUPAC Name 1,4-bis(dibromomethyl)benzene
SMILES C1=CC(=CC=C1C(Br)Br)C(Br)Br

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 421.75
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetrabromo-p-xylene
EINECSNumber (3)-0051
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.