Learn More
Invitrogen™ alpha Amylase 2B Monoclonal Antibody (OTI4B5)

Description
alpha Amylase 2B Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | alpha Amylase 2B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4B5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AMY2B |
| Gene Accession No. | P19961 |
| Gene Alias | 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase 2B; AMY2; Amy2-2; AMY2B; AMY3; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; OTTMUSG00000022462 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.