Learn More
CPC Scientific H-Ala-Pro-Arg-Leu-Arg-Phe-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific BACP001A
Encompass
SEQUENCE: H-Ala-Pro-Arg-Leu-Arg-Phe-Tyr-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: APRLRFYMOLECULAR FORMULA: C44H67N13O9MOLECULAR WEIGHT: 922.08STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [87549-54-0]SYNONYMS: RESEARCH AREA: HormonalREFERENCES:Pulst, S. et al. Neurosci. 27, 363 (1988)Rothman, B. et al. Proc. Natl. Acad. Sci. USA 80, 5753 (1983)SKU(s): BACP-001A
Catalog No. 50-210-8834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.