Learn More
Invitrogen™ TCR alpha/beta Monoclonal Antibody (WT31), PE, eBioscience™, Invitrogen™

Description
Description: This WT31 monoclonal antibody recognizes the human alpha beta T cell receptor (TCR) complexed with the CD3 epsilon chain. Composed of a 44-kDa alpha and a 37-kDa beta chain, TCR specificity is typically determined by V alpha, J alpha, V beta, D beta, and J beta gene rearrangement. The TCR is often associated with the CD3 complex, which is composed of at least five polypeptides. The TCR/CD3 complex recognizes and binds to major histocompatibility complex-bound antigen presented by antigen presenting cells such as B cells, dendritic cells, and macrophages. These events lead to T cell activation, resulting in cellular signaling and cytokine production. The WT31 monoclonal antibody has been reported to induce proliferation of resting T cells, as well as inhibit cytotoxic T cell activity. In addition, some antibodies against CD3 partially inhibit alpha beta TCR antibody staining using WT31; clone SK7 (anti-human CD3) is recommended for use in two-color experiments. Applications Reported: This WT31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This WT31 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Specifications
Specifications
| Antigen | TCR alpha/beta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | WT31 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | TRAC |
| Gene Accession No. | P01848, P04435, P0DSE1, P0DSE2 |
| Gene Alias | FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.