Learn More
Invitrogen™ alpha/beta Tubulin Monoclonal Antibody (TU-10)

Description
This antibody recognizes alpha, beta-tubulin heterodimer, a basic intracellular structural unit of microtubules. Alpha- and beta-tubulins form approximately 100 kDa tubulin heterodimer, a globular protein that polymerizes to form microtubules. Western Blot: Reducing conditions.
Specifications
Specifications
| Antigen | alpha/beta Tubulin |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | TU-10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 15mM sodium azide; pH 8 |
| Gene | TUBB |
| Gene Accession No. | A2AQ07, P02550, P07437, P68369, P68370, Q71U36, Q767L7, Q9H4B7 |
| Gene Alias | 2810484G07Rik; Alpha-tubulin 1; Alpha-tubulin 3; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; beta 5-tubulin; beta Ib tubulin; CDCBM6; CSCSC1; Detyrosinated tubulin alpha-1A chain; FLJ30169; H2 ALPHA; hum-a-tub1; hum-a-tub2; LIS3; M(beta)1; M40; OK/SW-cl.56; Talpha1 alpha-tubulin; Tuba1; Tuba-1; TUBA1A; TUBA3; TUBB; TUBB1; TUBB2A; TUBB5; Tubulin alpha; tubulin alpha 1a; tubulin alpha 3; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin beta; tubulin beta 1 class VI; tubulin beta chain; tubulin beta class I; tubulin beta-1 chain; Tubulin beta-5 chain; tubulin, alpha 1; tubulin, alpha 1A; tubulin, alpha, brain-specific; tubulin, beta 1 class VI; tubulin, beta 2A class IIa; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; α/β-tubulin |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.