Learn More
Description
- (-)-α-bisabolol is the major constituent of essential oil from leaves of hymenocrater yazdianus.(-)-α-bisabolol was used to investigate the leishmanicidal and cytotoxic activity of essential oil of vanillosmopsis arborea.α-bisabolol is active against primary acute leukemia cells, including bcr-abl(+) acute lymphoblastic leukemias
- It is the inhibitor of voltage-dependent ca(2+) channels in tracheal smooth muscle preparations of rat
- It also inhibits human and rat glioma
Specifications
Specifications
| CAS | 23089-26-1 |
| Molecular Formula | C15H26O |
| Refractive Index | n20/D 1.496 |
| Linear Formula | C15H26O |
| MDL Number | MFCD03412455 |
| Quantity | 25 mL |
| Synonym | (-)-alpha-Bisabolol; (-)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol; Levomenol |
| Molecular Weight (g/mol) | 222.37 |
| Percent Purity | ≥93% (GC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.