Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Bromo-gamma-butyrolactone, 97%

Catalog No. AAA1734414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1734414 25 g
AAA1734422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1734414 Supplier Thermo Scientific Chemicals Supplier No. A1734414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5061-21-2 | C4H5BrO2 | 164.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5061-21-2
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.99
MDL Number MFCD00005387
InChI Key LFJJGHGXHXXDFT-UHFFFAOYNA-N
Synonym 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
PubChem CID 95463
IUPAC Name 3-bromooxolan-2-one
SMILES BrC1CCOC1=O

Specifications

Density 1.994
Boiling Point 137°C to 139°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.508
Quantity 25 g
Beilstein 107800
Sensitivity Moisture sensitive
Formula Weight 164.99
Percent Purity 97%
Chemical Name or Material alpha-Bromo-gamma-butyrolactone
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 225-764-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.